Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cxcl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153576.2 | NP_705804.2 | 3 | 329 | ||
GenBank mRNA | AK143005.1 | - | 3 | 245 | ||
AY374329.1 | - | 3 | 232 | |||
AY598463.1 | - | 3 | 330 | |||
BC024561.1 | - | 3 | 329 | 5004999 |