Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153589.2 | NP_705817.2 | 3, 4 | 837 | ||
GenBank mRNA | AK044763.1 | - | 3, 4 | 838 | ||
BC033409.1 | - | 2, 3 | 432 | 5357763 | ||
BC060368.1 | - | 2, 3 | 441 | |||
FJ384096.1 | - | 3, 4 | 838 | |||
KC164759.2 | - | 3, 4 | 837 | |||
KC164761.2 | - | 2, 3 | 530 |