Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ano2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153589.2 | NP_705817.2 | 26 | 3857 | ||
GenBank mRNA | AK132944.1 | - | 18 | 2655 | ||
BC033409.1 | - | 25 | 3452 | 5357763 | ||
BC060368.1 | - | 26 | 3546 | |||
FJ384096.1 | - | 26 | 3858 | |||
KC164759.2 | - | 25 | 3845 | |||
KC164760.2 | - | 24 | 3746 | |||
KC164761.2 | - | 24 | 3538 | |||
KC164762.2 | - | 23 | 3439 |