Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Disp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170593.3 | NP_733481.1 | 8 | 6029 | ||
GenBank mRNA | AY150699.1 | - | 8 | 5830 |