Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RPL6 SIRNA Assays ›
Gene Symbol:
RPL6
Gene Name:
ribosomal protein L6
Gene Aliases:
L6, SHUJUN-2, TAXREB107, TXREB1
Chromosome Location:
Chr.12: 112405181 - 112418850 on Build GRCh38
UniGene ID:
Hs.528668
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
9349
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RPL6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000970.4
NP_000961.2
2
150
NM_001024662.2
NP_001019833.1
2
326
NM_001320137.1
NP_001307066.1
2
331
NM_001320138.1
NP_001307067.1
2
365
NM_001320139.1
NP_001307068.1
2
212
NM_001320140.1
NP_001307069.1
2
374
NM_001320141.1
NP_001307070.1
4
439
XM_017019781.1
XP_016875270.1
4
1474
XM_017019782.1
XP_016875271.1
4
635
GenBank mRNA
AF261087.1
-
1
117
AK298117.1
-
2
112
AK299470.1
-
3
80
AK308469.1
-
2
114
AK308680.1
-
2
144
AK311863.1
-
2
114
BC004138.2
-
1
126
2823733
BC013863.1
-
1
100
BC020679.1
-
1
114
4616668
BC022444.1
-
2
281
4286423
BC031009.1
-
1
114
4333761
BC032299.1
-
1
111
4994660
BC071912.1
-
1
103
6293404
BC104824.1
-
2
126
8143827
BC104826.1
-
2
126
8143829
BG722198.1
-
4
439
BP285025.1
-
2
150
BP308263.1
-
2
331
BU940020.1
-
149
CB140996.1
-
2
279
CD049618.1
-
2
296
CD640506.1
-
2
275
CD688935.1
-
2
381
D17554.1
-
2
116
HY016219.1
-
2
160
HY038019.1
-
4
439
X69391.1
-
2
121
Back To Top
More Information
Panther Classification:
Molecular Function -
ribosomal protein
Gene Ontology Categories:
Function(s)
Process(es)
ribosomal large subunit assembly
nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
cytoplasmic translation
regulation of transcription, DNA-templated
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
viral transcription
cell-cell adhesion
DNA binding
RNA binding
structural constituent of ribosome
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0