Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172086.2 | NP_742083.1 | 3 | 202 | ||
GenBank mRNA | AK002353.1 | - | 3 | 203 | ||
AK011017.1 | - | 3 | 203 | |||
AK012525.1 | - | 3 | 203 | |||
AK028204.1 | - | 3 | 203 | |||
AK146768.1 | - | 3 | 203 | |||
AK148453.1 | - | 3 | 203 | |||
AK150705.1 | - | 3 | 203 | |||
AK168282.1 | - | 3 | 206 | |||
BC030176.1 | - | 202 | ||||
BC046339.1 | - | 3 | 171 | 6478080 | ||
BC086909.1 | - | 3 | 206 | 30122780 |