Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crocc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172122.2 | NP_742120.2 | 5 | 686 | ||
GenBank mRNA | AF527975.1 | - | 5 | 686 | ||
BQ937768.1 | - | 2 | 212 |