Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pde8b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170669.1 | NP_001164140.1 | 2, 3 | 644 | ||
NM_172263.2 | NP_758467.2 | 2, 3 | 236 | |||
GenBank mRNA | AK036090.1 | - | 3, 4 | 564 | ||
AK043680.1 | - | 2, 3 | 637 | |||
AK045991.1 | - | 2, 3 | 645 | |||
AK046298.1 | - | 2, 3 | 660 | |||
AK083166.1 | - | 2, 3 | 282 | |||
BC032286.1 | - | 2, 3 | 248 | 5367055 | ||
BY244338.1 | - | 2, 3 | 270 | |||
CD806392.1 | - | 2, 3 | 209 | |||
EF644173.1 | - | 2, 3 | 98 |