Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc36a4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172289.4 | NP_758493.2 | 4, 5 | 381 | ||
GenBank mRNA | AF453746.1 | - | 4, 5 | 380 | ||
AK032074.1 | - | 4, 5 | 382 | |||
AK052277.1 | - | 4, 5 | 382 | |||
AK150167.1 | - | 4, 5 | 413 | |||
BC115964.1 | - | 4, 5 | 382 | 40057255 |