Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172125.2 | NP_742123.2 | 1 | 1383 | ||
GenBank mRNA | AK016558.1 | - | 1 | 1839 | ||
AK161264.1 | - | 1 | 1144 | |||
BC140979.1 | - | 1 | 1475 | 9055591 | ||
KF144404.1 | - | 1 | 168 | |||
KF144405.1 | - | 1 | 168 |