Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lcor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172154.4 | NP_742166.1 | 3 | 3596 | ||
GenBank mRNA | AB076079.1 | - | 8 | 1207 | ||
AK041090.1 | - | 3 | 3597 | |||
AK041621.1 | - | 8 | 1410 | |||
AK129442.1 | - | 6 | 1032 | |||
AK155007.1 | - | 8 | 1286 | |||
BC050068.1 | - | 3 | 578 | |||
BC090652.1 | - | 5 | 990 | 30657664 | ||
BC099938.1 | - | 8 | 1278 | 30085146 |