Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kdm1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172262.3 | NP_758466.1 | 3 | 313 | ||
GenBank mRNA | BC023917.1 | - | 3 | 254 | 5323227 | |
BY331331.1 | - | 3 | 273 | |||
CF531857.1 | - | 3 | 261 | |||
CF733387.1 | - | 2 | 76 |