Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snapc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290419.1 | NP_001277348.1 | 5 | 470 | ||
NM_172339.4 | NP_758842.1 | 4 | 351 | |||
GenBank mRNA | AK052107.1 | - | 4 | 378 | ||
AK077522.1 | - | 4 | 318 | |||
AK136996.1 | - | 4 | 328 | |||
BC018169.1 | - | 4 | 317 | |||
BC057031.1 | - | 5 | 436 | 6833968 |