Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Megf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162977.1 | NP_001156449.1 | 37 | 6234 | ||
GenBank mRNA | AK011141.1 | - | 1 | 303 | ||
AK173032.1 | - | 20 | 3713 | |||
BC031402.1 | - | 17 | 3272 | |||
BC039980.1 | - | 17 | 3283 |