Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172442.3 | NP_766030.3 | 3, 4 | 1775 | ||
GenBank mRNA | AK086624.1 | - | 2, 3 | 344 | ||
BC058647.1 | - | 3, 4 | 1233 | 6405881 | ||
BC069975.1 | - | 2, 3 | 315 | |||
CA328050.1 | - | 2, 3 | 212 |