Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zc3hav1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172467.3 | NP_766055.1 | 2, 3 | 540 | ||
GenBank mRNA | AK045287.1 | - | 2, 3 | 541 | ||
AK051064.1 | - | 2, 3 | 542 | |||
AK084475.1 | - | 2, 3 | 472 | |||
BC138180.1 | - | 2, 3 | 632 | 8861199 | ||
BC138181.1 | - | 2, 3 | 632 | 8861200 |