Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172410.2 | NP_765998.1 | 3 | 264 | ||
GenBank mRNA | AJ505051.1 | - | 2 | 200 | ||
AK010450.1 | - | 2 | 216 | |||
AK028864.1 | - | 3 | 265 | |||
AK129055.1 | - | 2 | 214 | |||
AK167105.1 | - | 2 | 215 | |||
AK169693.1 | - | 2 | 234 | |||
BC023140.1 | - | 2 | 210 | 3991335 |