Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nek11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172461.3 | NP_766049.2 | 13 | 1616 | ||
GenBank mRNA | AK030042.1 | - | 13 | 1575 | ||
AK030186.1 | - | 12 | 1492 | |||
BC119588.1 | - | 12 | 1450 | 40058889 | ||
BC119589.1 | - | 12 | 1450 | 40058890 |