Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sned1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172463.4 | NP_766051.4 | 31 | 4338 | ||
GenBank mRNA | AJ584850.1 | - | 31 | 4394 | ||
AK032805.1 | - | 9 | 1024 | |||
AK139277.1 | - | 7 | 450 | |||
BC037509.1 | - | 1 | 14 | |||
BC145886.1 | - | 31 | 4338 | 40131189 |