Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhlrc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172501.2 | NP_766089.1 | 2 | 215 | ||
GenBank mRNA | AK029465.1 | - | 2 | 210 | ||
AK033167.1 | - | 2 | 215 | |||
AK133986.1 | - | 165 | ||||
AK162824.1 | - | 2 | 255 | |||
AK220477.1 | - | 280 | ||||
BC087903.1 | - | 2 | 202 | 30250042 | ||
BC107356.1 | - | 2 | 203 | 40055875 |