Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145001.2 | NP_001138473.1 | 4 | 531 | ||
NM_001166167.1 | NP_001159639.1 | 3 | 313 | |||
NM_001166168.1 | NP_001159640.1 | 3 | 308 | |||
NM_001166169.1 | NP_001159641.1 | 3 | 317 | |||
NM_001166170.1 | NP_001159642.1 | 3 | 315 | |||
NM_001166171.1 | NP_001159643.1 | 4 | 480 | |||
NM_014397.5 | NP_055212.2 | 3 | 366 | |||
XM_005251664.1 | XP_005251721.1 | 3 | 372 | |||
XM_006716936.2 | XP_006716999.1 | 3 | 377 | |||
XM_017014217.1 | XP_016869706.1 | 4 | 540 | |||
GenBank mRNA | AB026289.1 | - | 3 | 297 | ||
AF087909.1 | - | 3 | 302 | |||
AK294614.1 | - | 3 | 317 | |||
AK313071.1 | - | 3 | 267 | |||
BC000101.2 | - | 3 | 312 | 3507262 | ||
BC004174.2 | - | 4 | 484 | 2958695 | ||
BC004209.2 | - | 4 | 484 | 2958695 | ||
BC012761.2 | - | 3 | 313 | 3640103 | ||
BM083872.1 | - | 3 | 313 | |||
BP265230.1 | - | 3 | 317 | |||
BP279761.1 | - | 3 | 308 | |||
CF994196.1 | - | 3 | 393 | |||
CR457091.1 | - | 2 | 147 | |||
CR542222.1 | - | 2 | 147 |