Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh5a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172532.3 | NP_766120.1 | 1, 2 | 419 | ||
GenBank mRNA | AK052703.1 | - | 1, 2 | 420 | ||
AK144030.1 | - | 1, 2 | 403 | |||
AK159485.1 | - | 1, 2 | 400 | |||
BC138703.1 | - | 1, 2 | 382 | 8861725 |