Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vezt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK031848.1 | - | 3 | 279 | ||
AK036017.1 | - | 3 | 227 | |||
AK041354.1 | - | 3 | 256 | |||
AK042540.1 | - | 3 | 221 | |||
AK163870.1 | - | 2 | 3105 | |||
BC056428.1 | - | 3 | 214 | 5716245 |