Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172578.2 | NP_766166.2 | 2 | 337 | ||
GenBank mRNA | AK035117.1 | - | 2 | 339 | ||
AK036395.1 | - | 2 | 347 | |||
AK053496.1 | - | 2 | 347 | |||
AK162657.1 | - | 2 | 361 | |||
BC042728.1 | - | 2 | 355 | |||
BC052175.1 | - | 2 | 337 | 6394068 |