Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dis3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001295.2 | NP_001001295.2 | 4 | 604 | ||
NM_001177784.1 | NP_001171255.1 | 4 | 565 | |||
NM_172519.3 | NP_766107.1 | 4 | 508 | |||
GenBank mRNA | AK029974.1 | - | 4 | 509 | ||
AK032451.1 | - | 4 | 513 | |||
AK035357.1 | - | 4 | 504 | |||
AK077031.1 | - | 4 | 568 | |||
AK150851.1 | - | 4 | 554 | |||
AK151780.1 | - | 4 | 554 | |||
AK153433.1 | - | 4 | 554 | |||
AK173310.1 | - | 4 | 538 | |||
BC056939.1 | - | 4 | 537 | 5708685 |