Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdr41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172590.3 | NP_766178.2 | 4 | 423 | ||
GenBank mRNA | AK046853.1 | - | 4 | 392 | ||
AK082348.1 | - | 4 | 424 | |||
AK149993.1 | - | 4 | 389 | |||
AK194741.1 | - | 3 | 90 | |||
BB650073.1 | - | 4 | 419 | |||
BC138095.1 | - | 4 | 329 | 8861114 | ||
BC145143.1 | - | 4 | 325 | |||
BC145144.1 | - | 4 | 329 | 9053695 |