Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncoa7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111267.2 | NP_001104737.1 | 6 | 1619 | ||
NM_172495.5 | NP_766083.3 | 16 | 3970 | |||
GenBank mRNA | AK042369.1 | - | 5 | 1904 | ||
AK128935.1 | - | 15 | 3793 | |||
AK171679.1 | - | 5 | 1464 | |||
BC076623.1 | - | 14 | 3576 | |||
BC080742.1 | - | 5 | 2725 | |||
EF602028.1 | - | 6 | 1619 |