Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mis18bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172578.2 | NP_766166.2 | 14 | 3852 | ||
GenBank mRNA | AK032698.1 | - | 12 | 2884 | ||
AK139165.1 | - | 3 | 825 | |||
BC021832.1 | - | 9 | 2360 | |||
BC052175.1 | - | 14 | 3852 | 6394068 |