Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFG3L1P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003226.1 | 10 | 1140 | |||
NR_003227.1 | 9 | 967 | ||||
NR_003228.1 | 10 | 1106 | ||||
GenBank mRNA | AF329691.1 | - | 9 | 891 | ||
AF329692.1 | - | 4 | 442 | |||
AJ001495.1 | - | 2 | 114 | |||
AK056488.1 | - | 10 | 1111 | |||
AK091293.1 | - | 10 | 1103 | |||
AK097044.1 | - | 9 | 954 | |||
AK126911.1 | - | 8 | 931 | |||
AK128352.1 | - | 3 | 806 | |||
AK128804.1 | - | 4 | 333 | |||
AL832984.1 | - | 4 | 364 | |||
BC021245.2 | - | 8 | 713 |