Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mier3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172593.3 | NP_766181.2 | 6, 7 | 697 | ||
GenBank mRNA | AK030747.1 | - | 6, 7 | 625 | ||
AK083929.1 | - | 5, 6 | 536 | |||
AK147427.1 | - | 6, 7 | 698 | |||
BC103780.1 | - | 5, 6 | 527 | 30108551 | ||
BC138127.1 | - | 5, 6 | 517 | 8861146 |