Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ambra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080754.1 | NP_001074223.1 | 2 | 471 | ||
NM_172669.3 | NP_766257.3 | 2 | 471 | |||
GenBank mRNA | AK037516.1 | - | 2 | 428 | ||
AK083596.1 | - | 2 | 428 | |||
AK084557.1 | - | 2 | 452 | |||
AK148887.1 | - | 2 | 463 | |||
AK155513.1 | - | 2 | 434 | |||
AK169280.1 | - | 2 | 424 | |||
BC032293.1 | - | 2 | 394 | |||
BC054082.1 | - | 2 | 403 |