Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txndc16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172597.2 | NP_766185.1 | 4 | 538 | ||
GenBank mRNA | AK017582.1 | - | 4 | 433 | ||
AK034025.1 | - | 4 | 527 | |||
AK039945.1 | - | 4 | 405 | |||
AK046466.1 | - | 4 | 539 | |||
AK164111.1 | - | 4 | 547 | |||
AK173153.1 | - | 1035 | ||||
AK215424.1 | - | 2 | 118 | |||
BC052488.1 | - | 4 | 414 | 6438726 | ||
BC060101.1 | - | 4 | 540 |