Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btbd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027060.1 | NP_081336.1 | 2, 3 | 347 | ||
NM_172618.2 | NP_766206.1 | 2, 3 | 347 | |||
GenBank mRNA | AK029578.1 | - | 2, 3 | 510 | ||
AK052664.1 | - | 2, 3 | 231 | |||
AK087881.1 | - | 2, 3 | 249 | |||
AK153973.1 | - | 2, 3 | 274 | |||
BC057897.1 | - | 2, 3 | 225 | 5031346 |