Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ldlrad4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172631.3 | NP_766219.2 | 6 | 1437 | ||
GenBank mRNA | AK040289.1 | - | 4 | 846 | ||
AK052338.1 | - | 6 | 1438 | |||
BC022716.1 | - | 3 | 558 | |||
BC096371.1 | - | 5 | 1198 | 3583946 | ||
BC131933.1 | - | 5 | 1045 | 40130210 | ||
BC131935.1 | - | 5 | 1045 | 40130212 |