Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hectd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163471.1 | NP_001156943.1 | 2 | 357 | ||
NM_172637.3 | NP_766225.2 | 2 | 357 | |||
GenBank mRNA | AK029549.1 | - | 2 | 217 | ||
AK041629.1 | - | 2 | 328 | |||
AK043646.1 | - | 2 | 318 | |||
AK079321.1 | - | 2 | 313 | |||
AK142777.1 | - | 2 | 357 | |||
BC137671.1 | - | 2 | 328 | 8860690 | ||
BC144815.1 | - | 2 | 328 | 9053363 |