Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Frmd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172673.3 | NP_766261.2 | 7, 8 | 873 | ||
GenBank mRNA | AK030883.1 | - | 7, 8 | 629 | ||
AK044270.1 | - | 7, 8 | 624 | |||
AK048865.1 | - | 7, 8 | 874 | |||
AK054484.1 | - | 7, 8 | 830 | |||
BC038623.1 | - | 7, 8 | 610 | |||
BC057549.1 | - | 4, 5 | 402 | 6416914 | ||
BC062889.1 | - | 7, 8 | 830 | 5698263 |