Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGDIB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001175.6 | NP_001166.3 | 2 | 362 | ||
NM_001321420.1 | NP_001308349.1 | 3 | 520 | |||
NM_001321421.1 | NP_001308350.1 | 2 | 241 | |||
NM_001321422.1 | NP_001308351.1 | 2 | 363 | |||
NM_001321423.1 | NP_001308352.1 | 2 | 229 | |||
GenBank mRNA | AA143425.1 | - | 2 | 170 | ||
AB451315.1 | - | 1 | 116 | |||
AB451445.1 | - | 1 | 116 | |||
AF498927.1 | - | 1 | 116 | |||
AK311807.1 | - | 2 | 192 | |||
AV685495.1 | - | 2 | 221 | |||
AV717900.1 | - | 2 | 240 | |||
BC009200.2 | - | 2 | 166 | 3621138 | ||
BE744996.1 | - | 2 | 227 | |||
BG390701.1 | - | 3 | 337 | |||
BI762648.1 | - | 2 | 361 | |||
BP355037.1 | - | 2 | 337 | |||
BY996333.1 | - | 2 | 226 | |||
CD385175.1 | - | 2 | 380 | |||
L07916.1 | - | 2 | 190 | |||
L20688.1 | - | 2 | 268 | |||
X69549.1 | - | 2 | 170 |