Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ino80c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172625.2 | NP_766213.1 | 5 | 2244 | ||
GenBank mRNA | AK083718.1 | - | 5 | 2245 | ||
BC056426.1 | - | 2 | 1947 |