Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kansl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172652.3 | NP_766240.1 | 21 | 4443 | ||
GenBank mRNA | AK030079.1 | - | 21 | 4444 | ||
AK129329.1 | - | 21 | 4307 | |||
BC023955.1 | - | 5 | 2294 |