Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172736.3 | NP_766324.1 | 6 | 813 | ||
GenBank mRNA | AK034310.1 | - | 6 | 818 | ||
AK170629.1 | - | 6 | 821 | |||
BC041775.1 | - | 6 | 785 | |||
BC042658.1 | - | 6 | 801 | 5365041 | ||
BC066768.1 | - | 6 | 804 | 6407146 |