Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172779.4 | NP_766367.1 | 2 | 521 | ||
GenBank mRNA | AK031945.1 | - | 2 | 513 | ||
AK083942.1 | - | 2 | 483 | |||
AK160627.1 | - | 2 | 481 | |||
BC051161.1 | - | 2 | 503 | 6466142 |