Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btnl9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172793.2 | NP_766381.1 | 2, 3 | 227 | ||
GenBank mRNA | AK080918.1 | - | 2, 3 | 210 | ||
AK084528.1 | - | 2, 3 | 228 | |||
AY177687.1 | - | 2, 3 | 194 | |||
BC025540.1 | - | 2, 3 | 205 | |||
BC052556.1 | - | 1, 2 | 140 | |||
BC150808.1 | - | 2, 3 | 184 | 9087719 | ||
BC151155.1 | - | 2, 3 | 184 | 9088043 |