Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc38a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172758.4 | NP_766346.1 | 5 | 840 | ||
GenBank mRNA | AK052548.1 | - | 5 | 841 | ||
AK171137.1 | - | 5 | 848 | |||
AK187529.1 | - | 1 | 68 | |||
AK218537.1 | - | 2 | 190 | |||
BC031853.1 | - | 3 | 715 | 4009949 |