Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163756.1 | NP_001157228.1 | 6, 7 | 984 | ||
NM_172759.3 | NP_766347.1 | 6, 7 | 984 | |||
GenBank mRNA | AK033563.1 | - | 6, 7 | 947 | ||
AK077248.1 | - | 6, 7 | 975 | |||
BC022148.1 | - | 2, 3 | 297 | |||
BC055062.1 | - | 6, 7 | 953 | 6528966 |