Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmo9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172844.2 | NP_766432.1 | 5, 6 | 707 | ||
GenBank mRNA | AK029219.1 | - | 5, 6 | 708 | ||
AK161224.1 | - | 5, 6 | 707 | |||
BC111885.1 | - | 3, 4 | 436 | 40060432 | ||
BC112405.1 | - | 5, 6 | 697 | 40060428 |