Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172814.3 | NP_766402.1 | 2, 3 | 207 | ||
GenBank mRNA | AK050512.1 | - | 2, 3 | 208 |