Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Peli3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172835.3 | NP_766423.3 | 7 | 1690 | ||
GenBank mRNA | AK044418.1 | - | 7 | 1637 | ||
AK135575.1 | - | 6 | 1538 | |||
BC028931.1 | - | 7 | 1655 | 5037308 | ||
BC057046.1 | - | 1 | 1912 |