Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wdfy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172882.3 | NP_766470.2 | 11 | 2025 | ||
GenBank mRNA | AK047077.1 | - | 11 | 2027 | ||
AY336569.1 | - | 11 | 2025 |