Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122.3 | NP_001113.2 | 4 | 491 | ||
NR_038064.1 | 3 | 578 | ||||
XM_017014259.1 | XP_016869748.1 | 3 | 303 | |||
GenBank mRNA | AF443203.1 | - | 3 | 281 | ||
AK296125.1 | - | 3 | 456 | |||
AK297920.1 | - | 4 | 403 | |||
AK312983.1 | - | 4 | 376 | |||
BC005127.2 | - | 4 | 357 | 3844174 | ||
BE311586.1 | - | 4 | 460 | |||
CN277218.1 | - | 3 | 580 | |||
CR536541.1 | - | 3 | 281 | |||
X97324.1 | - | 3 | 281 |