Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172931.3 | NP_766519.1 | 3, 4 | 368 | ||
GenBank mRNA | AB510446.1 | - | 4, 5 | 470 | ||
AB510447.1 | - | 970 | ||||
AK046229.1 | - | 398 | ||||
BC117805.2 | - | 3, 4 | 368 | 40092308 |